Assay Design



Gene Transcripts

Gene Symbol
WNT6
Entrez Gene ID
101111785
Gene Name
Wnt family member 6
Gene Aliases
-
Chromosome Location
Chr.2: 235648791 - 235661742 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965197.1 XP_027820998.1 1 - 2 84 83

View Full Product Details