Assay Design



Gene Transcripts

Gene Symbol
GOLT1A
Entrez Gene ID
101107979
Gene Name
golgi transport 1A
Gene Aliases
-
Chromosome Location
Chr.12: 1568983 - 1580856 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012186925.3 XP_012042315.2 2 - 3 227 96

View Full Product Details