Assay Design



Gene Transcripts

Gene Symbol
DNAJC24
Entrez Gene ID
101119247
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Gene Aliases
-
Chromosome Location
Chr.15: 67195922 - 67273591 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016366.4 XP_004016415.3 1 - 2 62 83
XM_012096147.3 XP_011951537.2 1 - 2 62 83

View Full Product Details