Assay Design



Gene Transcripts

Gene Symbol
SMAD6
Entrez Gene ID
101103068
Gene Name
SMAD family member 6
Gene Aliases
-
Chromosome Location
Chr.7: 14456246 - 14533727 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010255.4 XP_004010304.2 1 - 2 1933 76

View Full Product Details