Assay Design



Gene Transcripts

Gene Symbol
KLHL36
Entrez Gene ID
101103833
Gene Name
kelch like family member 36
Gene Aliases
-
Chromosome Location
Chr.14: 11511441 - 11527420 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977689.1 XP_027833490.1 4 - 5 1638 56
XM_027977690.1 XP_027833491.1 4 - 5 1729 56

View Full Product Details