Assay Design



Gene Transcripts

Gene Symbol
FGA
Entrez Gene ID
101102090
Gene Name
fibrinogen alpha chain
Gene Aliases
-
Chromosome Location
Chr.17: 3347762 - 3356083 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017183.3 XP_004017232.3 1 - 2 107 60

View Full Product Details