Assay Design



Gene Transcripts

Gene Symbol
SYNDIG1
Entrez Gene ID
101104331
Gene Name
synapse differentiation inducing 1
Gene Aliases
-
Chromosome Location
Chr.13: 43911285 - 44018937 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976372.1 XP_027832173.1 3 - 4 1017 70

View Full Product Details