Assay Design



Gene Transcripts

Gene Symbol
TSPAN19
Entrez Gene ID
101107267
Gene Name
tetraspanin 19
Gene Aliases
-
Chromosome Location
Chr.3: 129927687 - 129959150 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006232.4 XP_004006281.2 5 - 6 432 139
XM_004006233.4 XP_004006282.2 5 - 6 402 139
XM_012174038.3 XP_012029428.2 6 - 7 516 139

View Full Product Details