Assay Design



Gene Transcripts

Gene Symbol
MAP6
Entrez Gene ID
101109480
Gene Name
microtubule associated protein 6
Gene Aliases
-
Chromosome Location
Chr.15: 57868800 - 57949514 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979547.1 XP_027835348.1 2 - 3 1138 63

View Full Product Details