Assay Design



Gene Transcripts

Gene Symbol
CATSPERG
Entrez Gene ID
101111312
Gene Name
cation channel sperm associated auxiliary subunit gamma
Gene Aliases
-
Chromosome Location
Chr.14: 51027833 - 51060775 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978237.1 XP_027834038.1 27 - 28 5063 93

View Full Product Details