Assay Design



Gene Transcripts

Gene Symbol
PTPA
Entrez Gene ID
101105403
Gene Name
protein phosphatase 2 phosphatase activator
Gene Aliases
-
Chromosome Location
Chr.3: 7460706 - 7494483 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966316.1 XP_027822117.1 8 - 9 989 79

View Full Product Details