Assay Design



Gene Transcripts

Gene Symbol
PGLYRP1
Entrez Gene ID
101115395
Gene Name
peptidoglycan recognition protein 1
Gene Aliases
-
Chromosome Location
Chr.14: 57000426 - 57069818 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015325.4 XP_004015374.2 1 - 2 271 91

View Full Product Details