Assay Design



Gene Transcripts

Gene Symbol
PPIL1
Entrez Gene ID
101121978
Gene Name
peptidylprolyl isomerase like 1
Gene Aliases
-
Chromosome Location
Chr.20: 12415437 - 12438908 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018785.4 XP_004018834.1 1 - 2 338 93

View Full Product Details