Assay Design



Gene Transcripts

Gene Symbol
VGLL1
Entrez Gene ID
101117226
Gene Name
vestigial like family member 1
Gene Aliases
-
Chromosome Location
Chr.X: 107268025 - 107291338 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012107263.3 XP_011962653.2 3 - 4 850 81
XM_012107264.3 XP_011962654.2 3 - 4 861 81

View Full Product Details