Assay Design



Gene Transcripts

Gene Symbol
INA
Entrez Gene ID
101115747
Gene Name
internexin neuronal intermediate filament protein alpha
Gene Aliases
-
Chromosome Location
Chr.22: 26427149 - 26439680 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015103398.2 XP_014958884.2 1 - 2 1116 66

View Full Product Details