Assay Design



Gene Transcripts

Gene Symbol
GABARAPL2
Entrez Gene ID
101118469
Gene Name
GABA type A receptor associated protein like 2
Gene Aliases
-
Chromosome Location
Chr.14: 2410330 - 2421315 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015100375.2 XP_014955861.1 2 - 3 212 99

View Full Product Details