Assay Design



Gene Transcripts

Gene Symbol
LRRIQ4
Entrez Gene ID
101120037
Gene Name
leucine rich repeats and IQ motif containing 4
Gene Aliases
-
Chromosome Location
Chr.1: 236649232 - 236665274 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966187.1 XP_027821988.1 2 - 3 1189 82

View Full Product Details