Assay Design



Gene Transcripts

Gene Symbol
OTUB2
Entrez Gene ID
101114561
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 2
Gene Aliases
-
Chromosome Location
Chr.18: 59007075 - 59028236 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012098977.3 XP_011954367.1 2 - 3 177 108

View Full Product Details