Assay Design



Gene Transcripts

Gene Symbol
GRM6
Entrez Gene ID
443292
Gene Name
glutamate metabotropic receptor 6
Gene Aliases
MGLUR6
Chromosome Location
Chr.5: 2415864 - 2428160 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969215.1 XP_027825016.1 4 - 5 985 68

View Full Product Details