Assay Design



Gene Transcripts

Gene Symbol
PIRT
Entrez Gene ID
101103908
Gene Name
phosphoinositide interacting regulator of transient receptor potential channels
Gene Aliases
-
Chromosome Location
Chr.11: 33673782 - 33682268 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975467.1 XP_027831268.1 2 - 2 380 75

View Full Product Details