Assay Design



Gene Transcripts

Gene Symbol
DNAI2
Entrez Gene ID
101113084
Gene Name
dynein axonemal intermediate chain 2
Gene Aliases
-
Chromosome Location
Chr.11: 6144230 - 6176131 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012186587.3 XP_012041977.1 9 - 10 1368 55
XM_027974026.1 XP_027829827.1 9 - 10 1368 55
XM_027974027.1 XP_027829828.1 9 - 10 1365 55
XM_027974029.1 XP_027829830.1 9 - 10 1368 55

View Full Product Details