Assay Design



Gene Transcripts

Gene Symbol
CCDC90B
Entrez Gene ID
101105956
Gene Name
coiled-coil domain containing 90B
Gene Aliases
-
Chromosome Location
Chr.21: 14028963 - 14052241 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019415.4 XP_004019464.2 2 - 3 625 103

View Full Product Details