Assay Design



Gene Transcripts

Gene Symbol
CEACAM19
Entrez Gene ID
101111222
Gene Name
CEA cell adhesion molecule 19
Gene Aliases
-
Chromosome Location
Chr.14: 55941050 - 55959189 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978451.1 XP_027834252.1 4 - 5 886 83

View Full Product Details