Assay Design



Gene Transcripts

Gene Symbol
SPIN4
Entrez Gene ID
101109772
Gene Name
spindlin family member 4
Gene Aliases
-
Chromosome Location
Chr.X: 47790724 - 47794913 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012106557.3 XP_011961947.2 1 - 1 1094 96

View Full Product Details