Assay Design



Gene Transcripts

Gene Symbol
TMEM53
Entrez Gene ID
101109235
Gene Name
transmembrane protein 53
Gene Aliases
-
Chromosome Location
Chr.1: 19918867 - 19937069 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969131.1 XP_027824932.1 3 - 3 493 67

View Full Product Details