Assay Design



Gene Transcripts

Gene Symbol
POLR2D
Entrez Gene ID
101107845
Gene Name
RNA polymerase II subunit D
Gene Aliases
-
Chromosome Location
Chr.2: 127287697 - 127301188 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004488.4 XP_004004537.1 2 - 3 309 89

View Full Product Details