Assay Design



Gene Transcripts

Gene Symbol
LOC101103967
Entrez Gene ID
101103967
Gene Name
growth/differentiation factor 3
Gene Aliases
GDF3
Chromosome Location
Chr.3: 222158140 - 222164242 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006902.4 XP_004006951.2 1 - 2 266 62

View Full Product Details