Assay Design



Gene Transcripts

Gene Symbol
CAPNS1
Entrez Gene ID
678676
Gene Name
calpain small subunit 1
Gene Aliases
-
Chromosome Location
Chr.14: 49525890 - 49533013 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012190219.3 XP_012045609.1 9 - 10 827 64
GenBank mRNA DQ449954.1 - 6 - 7 471 64

View Full Product Details