Assay Design



Gene Transcripts

Gene Symbol
SIX2
Entrez Gene ID
101110829
Gene Name
SIX homeobox 2
Gene Aliases
-
Chromosome Location
Chr.3: 84590007 - 84594195 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005980.4 XP_004006029.3 1 - 2 807 97

View Full Product Details