Assay Design



Gene Transcripts

Gene Symbol
SNPH
Entrez Gene ID
101106451
Gene Name
syntaphilin
Gene Aliases
-
Chromosome Location
Chr.13: 62135513 - 62148340 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977030.1 XP_027832831.1 2 - 3 2321 108

View Full Product Details