Assay Design



Gene Transcripts

Gene Symbol
MAPRE3
Entrez Gene ID
101112123
Gene Name
microtubule associated protein RP/EB family member 3
Gene Aliases
-
Chromosome Location
Chr.3: 35591960 - 35646934 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015094261.2 XP_014949747.1 6 - 7 982 107
XM_015094262.2 XP_014949748.1 5 - 6 930 107
XM_027966593.1 XP_027822394.1 6 - 7 916 107

View Full Product Details