Assay Design



Gene Transcripts

Gene Symbol
ATP5MC3
Entrez Gene ID
101108285
Gene Name
ATP synthase membrane subunit c locus 3
Gene Aliases
ATP5G3
Chromosome Location
Chr.2: 145521312 - 145524449 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004569.3 XP_004004618.1 3 - 4 289 99

View Full Product Details