Assay Design



Gene Transcripts

Gene Symbol
ARHGEF38
Entrez Gene ID
101104491
Gene Name
Rho guanine nucleotide exchange factor 38
Gene Aliases
-
Chromosome Location
Chr.6: 23143793 - 23294949 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004009653.4 XP_004009702.2 9 - 10 1364 121

View Full Product Details