Assay Design



Gene Transcripts

Gene Symbol
LOC114117315
Entrez Gene ID
114117315
Gene Name
lymphocyte transmembrane adapter 1-like
Gene Aliases
-
Chromosome Location
Chr.12: 1088944 - 1093927 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976138.1 XP_027831939.1 4 - 4 808 106

Gene Symbol
LOC114117334
Entrez Gene ID
114117334
Gene Name
lymphocyte transmembrane adapter 1-like
Gene Aliases
-
Chromosome Location
Chr.12: 984353 - 999295 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976183.1 XP_027831984.1 5 - 5 976 106

View Full Product Details