Assay Design



Gene Transcripts

Gene Symbol
VHL
Entrez Gene ID
101122897
Gene Name
von Hippel-Lindau tumor suppressor
Gene Aliases
-
Chromosome Location
Chr.19: 17713634 - 17720266 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958083.1 XP_027813884.1 3 - 3 515 107

View Full Product Details