Assay Design



Gene Transcripts

Gene Symbol
P2RX4
Entrez Gene ID
100008720
Gene Name
purinergic receptor P2X 4
Gene Aliases
P2X4
Chromosome Location
Chr.21: 6348553 - 6365307 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002719744.3 XP_002719790.1 7 - 8 788 59
GenBank mRNA AF132031.1 - 3 - 4 247 59

View Full Product Details