Assay Design



Gene Transcripts

Gene Symbol
KCND1
Entrez Gene ID
100008836
Gene Name
potassium voltage-gated channel subfamily D member 1
Gene Aliases
-
Chromosome Location
Chr.X: 33608697 - 33617192 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002719905.3 XP_002719951.1 4 - 5 3181 59
GenBank mRNA AF493548.1 - 2 - 3 156 59

View Full Product Details