Assay Design



Gene Transcripts

Gene Symbol
GJC1
Entrez Gene ID
100009380
Gene Name
gap junction protein gamma 1
Gene Aliases
GJA7
Chromosome Location
Chr.19: 45521101 - 45522291 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199049.1 NP_001185978.1 1 - 1 792 93
GenBank mRNA DQ242487.1 - 1 - 1 552 93

View Full Product Details