Assay Design



Gene Transcripts

Gene Symbol
CD3E
Entrez Gene ID
100008616
Gene Name
CD3e molecule
Gene Aliases
-
Chromosome Location
Chr.Unknown: 376032 - 390796 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082001.1 NP_001075470.1 - 391 109
GenBank mRNA AB035151.1 - - 391 109

View Full Product Details