Assay Design



Gene Transcripts

Gene Symbol
HNRNPC
Entrez Gene ID
100008686
Gene Name
heterogeneous nuclear ribonucleoprotein C
Gene Aliases
HNRPC
Chromosome Location
Chr.17: 41457631 - 41512452 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082038.1 NP_001075507.1 4 - 5 482 113
XM_008268785.2 XP_008267007.1 5 - 6 601 113
XM_008268786.2 XP_008267008.1 4 - 5 573 113
XM_008268787.2 XP_008267009.1 3 - 4 527 113
XM_008268788.2 XP_008267010.1 5 - 6 601 113
XM_017347898.1 XP_017203387.1 5 - 6 601 113
GenBank mRNA AF061582.1 - 4 - 5 482 113

Gene Symbol
LOC100354921
Entrez Gene ID
100354921
Gene Name
heterogeneous nuclear ribonucleoprotein C
Gene Aliases
-
Chromosome Location
Chr.Unknown: 4627 - 6054 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008253032.2 XP_008251254.1 4 - 5 522 113

View Full Product Details