Assay Design



Gene Transcripts

Gene Symbol
LALBA
Entrez Gene ID
100008717
Gene Name
lactalbumin alpha
Gene Aliases
LYZG
Chromosome Location
Chr.4: 32948905 - 32988285 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082052.1 NP_001075521.1 3 - 4 400 149
XM_008256394.2 XP_008254616.1 4 - 5 467 149
XM_017341618.1 XP_017197107.1 4 - 5 433 149
GenBank mRNA AF124257.1 - 3 - 4 400 149

View Full Product Details