Assay Design



Gene Transcripts

Gene Symbol
ANKRD1
Entrez Gene ID
100008719
Gene Name
ankyrin repeat domain 1
Gene Aliases
-
Chromosome Location
Chr.18: 38909353 - 38917411 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082054.1 NP_001075523.1 7 - 8 968 68
GenBank mRNA AF131883.1 - 7 - 8 968 68

View Full Product Details