Assay Design



Gene Transcripts

Gene Symbol
KCNK1
Entrez Gene ID
100008962
Gene Name
potassium two pore domain channel subfamily K member 1
Gene Aliases
rabKCNK1
Chromosome Location
Chr.16: 34452022 - 34495175 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082180.1 NP_001075649.1 1 - 2 357 68
GenBank mRNA AF004695.1 - 1 - 2 354 68
AY752982.1 - 1 - 2 357 68

View Full Product Details