Assay Design



Gene Transcripts

Gene Symbol
WNT11
Entrez Gene ID
100009030
Gene Name
Wnt family member 11
Gene Aliases
WNT-11
Chromosome Location
Chr.1: 140610815 - 140630036 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082224.1 NP_001075693.1 1 - 2 87 52
XM_008258421.2 XP_008256643.1 2 - 3 505 52
GenBank mRNA DQ388601.1 - 1 - 2 87 52

View Full Product Details