Assay Design



Gene Transcripts

Gene Symbol
C8A
Entrez Gene ID
100009076
Gene Name
complement C8 alpha chain
Gene Aliases
-
Chromosome Location
Chr.13: 109972216 - 110036188 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082255.1 NP_001075724.1 9 - 10 1401 110
XM_008263970.2 XP_008262192.1 10 - 11 1565 110
XM_017345487.1 XP_017200976.1 10 - 11 1565 110
GenBank mRNA L26981.1 - 9 - 10 1401 110

View Full Product Details