Assay Design



Gene Transcripts

Gene Symbol
C8A
Entrez Gene ID
100009076
Gene Name
complement C8 alpha chain
Gene Aliases
-
Chromosome Location
Chr.13: 109972216 - 110036188 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082255.1 NP_001075724.1 10 - 11 1629 55
XM_008263970.2 XP_008262192.1 11 - 12 1793 55
GenBank mRNA L26981.1 - 10 - 11 1629 55

View Full Product Details