Assay Design



Gene Transcripts

Gene Symbol
VTN
Entrez Gene ID
100009128
Gene Name
vitronectin
Gene Aliases
VN
Chromosome Location
Chr.19: 19745310 - 19748086 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082292.1 NP_001075761.1 7 - 8 1341 64
GenBank mRNA M55442.1 - 7 - 8 1341 64

View Full Product Details