Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082315.1 | NP_001075784.1 | 9 - 10 | 1189 | 106 |
XM_008248341.2 | XP_008246563.1 | 9 - 10 | 1251 | 106 | |
XM_008253164.2 | XP_008251386.1 | 8 - 9 | 1151 | 106 | |
GenBank mRNA | M95815.1 | - | 9 - 10 | 1189 | 106 |