Assay Design



Gene Transcripts

Gene Symbol
SLC5A6
Entrez Gene ID
100009326
Gene Name
solute carrier family 5 member 6
Gene Aliases
SMVT
Chromosome Location
Chr.2: 158859474 - 158872872 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082637.1 NP_001076106.1 4 - 5 856 77
GenBank mRNA AF080067.1 - 4 - 5 856 77

View Full Product Details