Assay Design



Gene Transcripts

Gene Symbol
SPA17
Entrez Gene ID
100009312
Gene Name
sperm autoantigenic protein 17
Gene Aliases
-
Chromosome Location
Chr.1: 92552405 - 92567388 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082626.1 NP_001076095.1 - 759 91
GenBank mRNA Z20655.1 - - 759 91

View Full Product Details